Learn More
Apexbio Technology LLC Rink Amide Linker 145069-56-3 100g
SDP
Catalog No. 5000094980
Encompass_Preferred
Rink Amide Linker is a synthetic linker commonly used in solid-phase peptide synthesis to generate peptides with C-terminal amide functionalities It enables controlled cleavage through acid-labile benzyl ester bonds facilitating the selective release of amide-terminated peptides upon treatment with acidic reagents Based on these chemical properties Rink Amide Linker holds research potential in peptide drug analog development structure-activity relationship (SAR) studies immunology-related peptide research and the preparation of biologically relevant amino-terminal modified peptide libraries
Catalog No. 50-000-94980 Supplier Apexbio Technology LLC Supplier No. A7042100000
Add to Cart
Add to Cart
Rink Amide Linker is a synthetic linker commonly used in solid-phase peptide synthesis to generate peptides with C-terminal amide functionalities It enables controlled cleavage through acid-labile benzyl ester bonds facilitating the selective release of amide-terminated peptides upon treatment with acidic reagents Based on these chemical properties Rink Amide Linker holds research potential in peptide drug analog development structure-activity relationship (SAR) studies immunology-related peptide research and the preparation of biologically relevant amino-terminal modified peptide libraries