Learn More
Chem-Impex International, Inc. RINK AMIDE-MBHA RESIN 0.3 25G
Catalog No. NC2480588
Encompass_Preferred
Rink amide-MBHA resin (0.3 - 0.6 mmol/g, 100 - 200 mesh), 25g,
Catalog No. NC2480588 Supplier Chem-Impex International, Inc. Supplier No. 3329725G
Rink amide-MBHA resin (0.3 - 0.6 mmol/g, 100 - 200 mesh), 25g,