Learn More
Carbosynth LLC. Rink-Amide-MBHA Resin (100-200 mesh) 1% DVB, 5g, 0.3-0.6 meq/g
SDP
Catalog No. 501686901
Encompass_Preferred
Synonyms: 4-(2',4'-Dimethoxyphenyl-Fmoc-Aminomethyl) Phenoxyacetyl-MBHA Resin, Application: Unsubstituted Resins for Solid Phase Synthesis, Storage Temperature: 2-10°C
Catalog No. 50-168-6901 Supplier Carbosynth LLC. Supplier No. RFR1063PI5G
Synonyms: 4-(2',4'-Dimethoxyphenyl-Fmoc-Aminomethyl) Phenoxyacetyl-MBHA Resin, Application: Unsubstituted Resins for Solid Phase Synthesis, Storage Temperature: 2-10°C