Learn More
Chem-Impex International, Inc. Rink amide resin (0.3 - 0.6 mmol/g, 100 - 200 mesh) | MFCD00801462 | 25G
Catalog No. NC2146560
Encompass_Preferred
Rink amide resin, (0.3-0.6 meq/g, 100-200 mesh, 25g
This item is not returnable. View return policy
Catalog No. NC2146560 Supplier Chem-Impex International, Inc. Supplier No. 1266225G
Rink amide resin, (0.3-0.6 meq/g, 100-200 mesh, 25g