Learn More
Sigma Aldrich Fine Chemicals Biosciences Rink Amide resin 100-200 m
Catalog No. 502901677
Encompass
An excellent support for the fmoc SPPS of peptide amides.This resin is more acid sensitive than Rink Amide AM and Rink Amide MBHA resins. Cleavage with high concentrations of TfA can lead to the breakdown of the linker with the concomitant formation of by-products that can not be removed by simple washes. These problems appear to be minimized through the use of low TfA concentrations or by the addition of trialkylsilanes to the cleavage mixture.The product number for this product was previously 01-64-0013.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-290-1677 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8550010005
An excellent support for the fmoc SPPS of peptide amides.This resin is more acid sensitive than Rink Amide AM and Rink Amide MBHA resins. Cleavage with high concentrations of TfA can lead to the breakdown of the linker with the concomitant formation of by-products that can not be removed by simple washes. These problems appear to be minimized through the use of low TfA concentrations or by the addition of trialkylsilanes to the cleavage mixture.The product number for this product was previously 01-64-0013.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.