Learn More
Chem-Impex International, Inc. Rink amide resin (0.3 - 0.6 mmol/g, 100 - 200 mesh) | MFCD00801462 | 5G
Catalog No. 50506906
Encompass_Preferred
Rink amide resin (0.3 - 0.6 mmol/g, 100 - 200 mesh), MFCD00801462, 5G
Catalog No. 50-506-906 Supplier Chem-Impex International, Inc. Supplier No. 126625G
Rink amide resin (0.3 - 0.6 mmol/g, 100 - 200 mesh), MFCD00801462, 5G