Learn More
Chem-Impex International, Inc. Rink amide resin (0.3 - 0.8 mmol/g, 200 - 400 mesh) | MFCD00801462 | 5G
Catalog No. 50505595
Encompass_Preferred
Rink amide resin (0.3 - 0.8 mmol/g, 200 - 400 mesh), MFCD00801462, 5G
Catalog No. 50-505-595 Supplier Chem-Impex International, Inc. Supplier No. 106195G
Rink amide resin (0.3 - 0.8 mmol/g, 200 - 400 mesh), MFCD00801462, 5G