Learn More
CEM Corporation Rink Amide ProTide Resin (LL), 25 g
SDP
Catalog No. 501877766
Encompass
ProTide resins contain an ideal PEG and polystyrene core leading to an optimized SPPS environment. Unmatched performance for routine synthesis of long (> 30 amino acids) or difficult peptide amides. Can be used in place of PAL or other Rink Amide resins.
Catalog No. 50-187-7766 Supplier CEM Corporation Supplier No. R002B5
ProTide resins contain an ideal PEG and polystyrene core leading to an optimized SPPS environment. Unmatched performance for routine synthesis of long (> 30 amino acids) or difficult peptide amides. Can be used in place of PAL or other Rink Amide resins.