Learn More
MilliporeSigma Fluid A in bottle with screw cap and septum 600ml, 4 bottles
Catalog No. NC0385495
Encompass
The Fluid A can be used in combination with the Steritest™ sterility testing system or for bioburden testing to rinse membranes and dilute/dissolve samples. 600ml
Catalog No. NC0385495 Supplier MilliporeSigma Supplier No. STBMRFA64
The Fluid A can be used in combination with the Steritest™ sterility testing system or for bioburden testing to rinse membranes and dilute/dissolve samples. 600ml