Learn More
MilliporeSigma RINSE FUILD A 12/PK 100ML
Supplier:  MilliporeSigma STBMRFA12
Encompass
Description Catalogue NumberSTBMRFa12 DescriptionFluid a in 100ml bottle with screw cap and septum OverviewThe Fluid a is suitable as a general rinse buffer, and compatible with most samples. Excellent for dissolving or diluting samples, reconstituting commercial microorganisms, or as a transport medium for microorganisms. Product Information Components Screw cap with septum applicable Device Steritesta, c Type of Test Sterility Testing applications applicationThe Fluid a can be used in combination with the Steritesta, c sterility testing system or for bioburden testing to rinse membranes and dilute/dissolve samples. Key applications Pharmaceutical Sterility Testing USP 71 Sterility Testing Biological Information Pre-filled agar mediaFluid a QC Organisms S. aureus (aTCC 6538) C. sporogenes (aTCC 11437) a. niger (aTCC 16404) C. albicans (aTCC 10231) P. aeruginosa (aTCC 9027) B. subtilis (aTCC 6633) Target Microorganisms Rinse Buffers and additives Media ColorClear with n
Catalog No. NC9486346