Learn More
ELITechGroup Aerospray Hematology Stat Reagent A, Rinse, 1Gal
SDP
Catalog No. NC9894817
Encompass
Aqueous Eosin Y Stain; Liquid, Orange; ~ 6 pH; 1 gal.; Hematology Reagent A Rinse; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light
Catalog No. NC9894817 Supplier ELITechGroup Supplier No. SS035AG
Aqueous Eosin Y Stain; Liquid, Orange; ~ 6 pH; 1 gal.; Hematology Reagent A Rinse; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light