Learn More
Ethos Biosciences Hematology Rinse Solution, pH 7.0, 5gal, Ethos
Catalog No. 5000574324
Encompass
Hematology Rinse Solution pH 7.0 is a buffered solution designed for use in hematology staining procedures. Commonly used as a rinse or dilution step in Romanowsky-type staining protocols, it helps remove excess stain while maintaining optimal pH for accurate cellular differentiation. The solution supports consistent staining quality, clear nuclear and cytoplasmic detail, and reliable microscopic evaluation. Ideal for peripheral blood smears and related hematology workflows, it is suitable for routine diagnostic and research applications.
Catalog No. 50-005-74324 Supplier Ethos Biosciences Supplier No. 55725G
Hematology Rinse Solution pH 7.0 is a buffered solution designed for use in hematology staining procedures. Commonly used as a rinse or dilution step in Romanowsky-type staining protocols, it helps remove excess stain while maintaining optimal pH for accurate cellular differentiation. The solution supports consistent staining quality, clear nuclear and cytoplasmic detail, and reliable microscopic evaluation. Ideal for peripheral blood smears and related hematology workflows, it is suitable for routine diagnostic and research applications.