Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rinse Solution, High Purity Water for Rinsing Electrodes and other Laboratory Instruments, Ricca Chemical
SDP

Catalog No. 664820 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
20 x 20 mL
200 x 20 mL
Packaging:
Natural Poly Bottle
Pouches
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6648-20 20 x 20 mL Pouches
6648-16 500 mL Natural Poly Bottle
6648-20B 200 x 20 mL Pouches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6648-20 Supplier Ricca Chemical Company Supplier No. 664820
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Percent Purity 100%
Grade Laboratory
Packaging Pouches
Quantity 20 x 20 mL
Physical Form Liquid
Chemical Name or Material Rinse Solution, High Purity Water for Rinsing Electrodes and other Laboratory Instruments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.