Learn More
Andwin Scientific RINZL COVERSLIP 24X50 1000/PK
SDP
Catalog No. 50949451
Encompass_Preferred
Constructed with clear vinyl plastic, the 24x50mm Rinzl Plastic Coverslips are resistant to breakage. This product also removes the risk of scratching or cutting the fingers of the users. These coverslips are ideal for use in biological and medical laboratories. This is sold in a pack of 1000.
Catalog No. 50-949-451 Supplier Andwin Scientific Supplier No. 7226150
Constructed with clear vinyl plastic, the 24x50mm Rinzl Plastic Coverslips are resistant to breakage. This product also removes the risk of scratching or cutting the fingers of the users. These coverslips are ideal for use in biological and medical laboratories. This is sold in a pack of 1000.