Learn More
Cayman Chemical Riociguat-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2749393-44-8, 1 mg
SDP
Catalog No. 502932494
Encompass_Preferred
An internal standard for the quantification of riociguat by GC- or LC-MS
Catalog No. 50-293-2494 Supplier Cayman Chemical Supplier No. 253141 MG
An internal standard for the quantification of riociguat by GC- or LC-MS