Learn More
Cayman Chemical RIocIguat-13C-d3 500ug
SDP
Catalog No. 502932527
Encompass_Preferred
An internal standard for the quantification of riociguat by GC- or LC-MS
Catalog No. 50-293-2527 Supplier Cayman Chemical Supplier No. 25314500 UG
An internal standard for the quantification of riociguat by GC- or LC-MS