Learn More
Apexbio Technology LLC Riociguat 625115-55-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064369
Encompass_Preferred
Riociguat (CAS 625115-55-1) chemically known as BAY 63-2521 is a small-molecule stimulator of soluble guanylate cyclase (sGC) It directly activates sGC independently and also enhances the enzyme s sensitivity to nitric oxide (NO) leading to elevated cyclic guanosine monophosphate (cGMP) levels Elevated cGMP induces smooth muscle relaxation and vasodilation Riociguat has been examined clinically for potential therapeutic roles in chronic thromboembolic pulmonary hypertension (CTEPH) and pulmonary arterial hypertension (PAH)
Catalog No. 50-000-64369 Supplier Apexbio Technology LLC Supplier No. A37675.1
 Restricted Item
Add to Cart
Add to Cart
Riociguat (CAS 625115-55-1) chemically known as BAY 63-2521 is a small-molecule stimulator of soluble guanylate cyclase (sGC) It directly activates sGC independently and also enhances the enzyme s sensitivity to nitric oxide (NO) leading to elevated cyclic guanosine monophosphate (cGMP) levels Elevated cGMP induces smooth muscle relaxation and vasodilation Riociguat has been examined clinically for potential therapeutic roles in chronic thromboembolic pulmonary hypertension (CTEPH) and pulmonary arterial hypertension (PAH)