Learn More
Aviva Systems Biology Corp RIOK2 Antibody - middle region (ARP57210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIOK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RIOK2. Alternate names: FLJ11159, RIO2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-280