Learn More
Aviva Systems Biology Corp RIOK3 Antibody - C-terminal region (ARP42152_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42152_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIOK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RIOK3. Alternate names: DKFZp779L1370, SUDDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58125