Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RiOs™ 3, 5, 8 Pure Water Purification System

Catalog No. p-4373396 Shop All MilliporeSigma Products
Change view
Click to view available options
Flow Rate:
8
Weight (Metric):
5.9 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Weight (Metric)
ZR0S0P8US 8 5.9 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZR0S0P8US Supplier MilliporeSigma™ Supplier No. ZR0S0P8US
Only null left
Add to Cart
Add to Cart
Request a Quote

Unique, all-in-one purification cartridges include pretreatment and high-performance reverse osmosis (RO) membranes that generate reliable, affordable laboratory-grade water for Type III applications or as pretreatment to ultrapure systems from normal tap

Produce flow rates adapted to your daily usage—3, 5, or 8L/hour

  • Small footprint, easy-to-install system can be placed wherever needed, on the benchtop or wall
  • Produce reliable and affordable Type 3 water from tap
  • User-friendly, backlit display provides easily accessible information on system status (conductivity, temperature values, water level in reservoir and alarms) at a glance; concise Quick Reference Guide is handy guide for daily operation
  • Convenient reservoir storage: RiOs 3 offers an integrated 6L reservoir, RiOs 5 and RiOs 8 can connect to external 30L or 60L reservoirs
  • Single SmartPak™ RO cartridge with integrated RO membrane enables easy and rapid replacement
  • Efficient removal of >96% of ionic contaminants and ≥99% of other contaminants (organics, bacteria, particles)
  • Simple, low-level self-maintenance—just change a single cartridge once or twice a year

Feedwater for ultrapure water systems (Synergy™ or Milli-Q™ systems), humidifiers, autoclaves or glassware washers; manual cleaning and glassware rinsing; buffer preparation

Order Info

19.7 x 11.4 x 13 in. (50 x 29 x 33 cm). 100-250V 50-60Hz. Tap pressure: 0.5 to 6 bar. Feedwater connection: 0.5 in. Gaz M.

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Single SmartPak™ reverse osmosis
Contaminant Rejection (Minimum Ionic) Greater than96%
Contaminant Rejection (Organic) <99%
Feedwater Source Tap
Flow Rate 8
Product Water Type Type 3
Display Backlit LCD
For Use With (Application) Removes more than 96% ionic contaminants, 99% other contaminants
Width (English) 11.4 in.
Width (Metric) 29 cm
Height (English) 19.7 in.
Height (Metric) 50 cm
Depth (English) 13 in.
Depth (Metric) 33 cm
Weight (English) 13.2 lb.
Weight (Metric) 5.9 kg
Electrical Requirements 100/250 V 50/60 Hz
Voltage 100/250 V
Frequency 50/60 Hz
Product Type RiOs Pure Water Purification System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.