Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RIP2 Kinase Inhibitor 3 | 1398053-50-3 | 99.76% | 388.48 | 10 MG
SDP

Catalog No. 5000438761
Encompass_Preferred

RIP2 Kinase Inhibitor 3 is a highly potent and selective inhibitor of receptor interacting protein-2 (RIP2) Kinase with an IC50 of 1 nM.

  • Highly potent and selective inhibitor of RIP2 Kinase.
  • IC50: 1 nM (RIP2).
  • Purity: 99.76%.
  • Appearance: Solid, light yellow to yellow.

Catalog No. 50-004-38761 Supplier Medchemexpress LLC Supplier No. HY11290710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RIP2 Kinase Inhibitor 3 is a highly potent and selective inhibitor of receptor interacting protein-2 (RIP2) Kinase with an IC50 of 1 nM.

  • Highly potent and selective inhibitor of RIP2 Kinase.
  • IC50: 1 nM (RIP2).
  • Purity: 99.76%.
  • Appearance: Solid, light yellow to yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.