Learn More
Enzo Life Sciences RIP 3 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905242100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse RIP3. UniProt: Q9QZL0. Species Reactivity: Mouse, Rat. Applications: IHC (PS), WB. Recommended dilutions: Immunohistochemistry (paraffin sections, 5µg/ml). Western Blot (0.5-1.0µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-658-304