Learn More
Enzo Life Sciences RIP3 (human) monoclonal antibody (Rippy-3) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804155C100
Encompass_Preferred
Clone: Rippy-3. Isotype: IgG1. Immunogen: Recombinant human RIP3 (receptor-interacting protein 3). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Applications: WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9802