Learn More
Medchemexpress LLC Ripa-56 | 1956370-21-0 | 99.9% | 221.30 g/mol | C13H19NO2 | 25 MG
SDP
Catalog No. 5000213082
Encompass_Preferred
RIPA-56 is a potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) used in biochemical and cellular research to study necroptosis and inflammatory signaling. The compound is supplied at high purity and is suitable for in vitro assays, available as a solid or as a DMSO stock solution.
  • Potent and selective RIP1 kinase inhibitor with low-nanomolar activity.
  • High purity suitable for biochemical and cellular assays.
  • Metabolically stable for in vitro pharmacology studies.
  • Available as solid and as DMSO stock solution for flexible dosing.
  • Supplied in small milligram quantities for research-scale experiments.
Catalog No. 50-002-13082 Supplier Medchemexpress LLC Supplier No. HY10103225MG
RIPA-56 is a potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) used in biochemical and cellular research to study necroptosis and inflammatory signaling. The compound is supplied at high purity and is suitable for in vitro assays, available as a solid or as a DMSO stock solution.
  • Potent and selective RIP1 kinase inhibitor with low-nanomolar activity.
  • High purity suitable for biochemical and cellular assays.
  • Metabolically stable for in vitro pharmacology studies.
  • Available as solid and as DMSO stock solution for flexible dosing.
  • Supplied in small milligram quantities for research-scale experiments.