Learn More
Biotang Inc RIPA Buffer with EDTA 5 mM & EGTA 1 mM, 250 ml
Catalog No. 50152127 Shop All BIOTANG Inc Products
Encompass_Preferred
RIPA Buffer with EDTA 5 mM & EGTA 1 mM, 250 ml.
Catalog No. 50-152-127 Supplier Biotang Inc Supplier No. BTBB608
RIPA Buffer with EDTA 5 mM & EGTA 1 mM, 250 ml.