Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals RIPA buffer

Catalog No. AAJ63306AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63306AK 250 mL
AAJ63306AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63306AK Supplier Thermo Scientific Chemicals Supplier No. J63306AK
Add to Cart

missing translation for 'validMainframeMsg'

Sorbitan monooleate, CAS # 1338-43-8, is a synthetic fatty acid ester showing emulsifying and dispersing activities.

RIPA is used as a high quality, ready to use mammalian cell and tissue lysis buffer with protease inhibitors included, is used to lyse cells and tissue, for radio immunoprecipitation assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Sorbitan monooleate is derived from sorbitol and is a synthetic fatty acid ester that presents emulsifying, dispersing, and wetting activities

Application

• Sorbitan monooleate is used in yeast laboratory strains to protect them from excess drying and also helps rehydrate yeast cells

• It can be used as a dispersant and as a corrosion inhibitor

• Sorbitan monooleate has important uses as an excipient and intravenous osmotic diuretic

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 250 mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.