Learn More
Alkali Scientific RIPA Buffer 2X Solution, 250 Milliliters
SDP
Catalog No. 5000661899
Encompass_Preferred
RIPA Buffer 2X Solution is a highly effective reagent supplied in a 250mL bottle for protein extraction, cell lysis, and sample preparation. The buffer contains a mixture of salts, detergents, and protease inhibitors that facilitate the breakdown of cell membranes and the solubilization of proteins. It is particularly useful for preparing samples for applications such as Western blotting, immunoprecipitation, and enzyme assays. The 2X concentration allows for easy dilution to the desired working strength, ensuring optimal performance. RIPA Buffer is widely used in molecular biology, cell biology, and biochemistry laboratories for the extraction of total cellular proteins from various tissues and cell lines.
Catalog No. 50-006-61899 Supplier Alkali Scientific Supplier No. RIP002CP
RIPA Buffer 2X Solution is a highly effective reagent supplied in a 250mL bottle for protein extraction, cell lysis, and sample preparation. The buffer contains a mixture of salts, detergents, and protease inhibitors that facilitate the breakdown of cell membranes and the solubilization of proteins. It is particularly useful for preparing samples for applications such as Western blotting, immunoprecipitation, and enzyme assays. The 2X concentration allows for easy dilution to the desired working strength, ensuring optimal performance. RIPA Buffer is widely used in molecular biology, cell biology, and biochemistry laboratories for the extraction of total cellular proteins from various tissues and cell lines.