Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals RIPA buffer (2X)

Catalog No. AAJ63324EQE
Change view
Click to view available options
Quantity:
125mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63324EQE 125mL
AAJ63324AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63324EQE Supplier Thermo Scientific Chemicals Supplier No. J63324EQE
Add to Cart

missing translation for 'validMainframeMsg'

RIPA is used as a high quality, ready to use mammalian cell and tissue lysis buffer. If protease inhibitors are included after receipt, it can be used to lyse cells and tissue, for radio immunoprecipitation assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 125mL
Format Liquid
Product Type Cell Lysis Reagent
Concentration 2X
Content And Storage 2° to 8°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.