Learn More
Sigma Aldrich Fine Chemicals Biosciences RIPA Buffer
Catalog No. 502900017
Encompass
Radioimmunoprecipitation assay buffer (RIPA buffer) is a lysis buffer used for rapid efficient cell lysis and solubilization of proteins from both adherent and suspension cultured mammalian cells.
Catalog No. 50-290-0017 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R027850ML
Radioimmunoprecipitation assay buffer (RIPA buffer) is a lysis buffer used for rapid efficient cell lysis and solubilization of proteins from both adherent and suspension cultured mammalian cells.