Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals RIPA buffer (5X)

Catalog No. AAJ62524AE
Change view
Click to view available options
Quantity:
50mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62524AE 100mL
AAJ62524AD 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62524AE Supplier Thermo Scientific Chemicals Supplier No. J62524AE
Add to Cart
Edge

missing translation for 'validMainframeMsg'

RIPA buffer, radio immunoprecipitation assay buffer, is used for whole cell isolation of proteins from tissues and cell culture.

RIPA buffer is used to lyse cells and tissues. It is used to lyse cultured mammalian cells from both plated cells and cells pelleted from suspension cultures. This buffer enables protein extraction from cytoplasmic, membrane and and is particularly useful for disruption of nuclear membranes in the preparation of nuclear extracts. It is compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification. This product is ideal for general cell-tissue lysis and sub cellular organelle or molecular fractionation, including immunoprecipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• RIPA can be used for isolation of cytoplasmic, membrane, nuclear, and mitochondrial proteins
• Protease inhibitors can be added to RIPA buffer to assist in protein isolation

Application

• RIPA buffer is suitable for use in protein isolation from culture cells or homogenized tissues
• Downstream applications of RIPA buffer include ELISA, western blot, immunoprecipitation, and SDS-PAGE

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mL
Format Liquid
Product Type Cell Lysis Reagent
Concentration 5X
Content And Storage 2° to 8°C
What are the concentrations of the components in RIPA buffer (5X) (Cat. No. J62524.xx)?

RIPA buffer (5X) contains 250 mM Tris-HCl (pH 7.4), 750 mM NaCl, 5% NP-40, 2.5% sodium deoxycholate, and 0.5% SDS.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.