Learn More
Teknova RIPA Buffer Solution. 500 mL, Sterile. 4 pack.
SDP
Catalog No. 502387607
Encompass
150mM Sodium Chloride, 1.0% Triton X-100, 1.0% Sodium Deoxycholate, 0.1% Sodium Dodecyl Sulfate, 50mM Tris-HCl,2mM Ethylenediaminetetraacetic acid (EDTA)
Catalog No. 50-238-7607 Supplier Teknova Supplier No. R37924PK
150mM Sodium Chloride, 1.0% Triton X-100, 1.0% Sodium Deoxycholate, 0.1% Sodium Dodecyl Sulfate, 50mM Tris-HCl,2mM Ethylenediaminetetraacetic acid (EDTA)