Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals RIPA buffer with Triton∣r X-100 (5X)

Catalog No. AAJ62885AE
Change view
Click to view available options
Quantity:
50mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62885AE 100mL
AAJ62885AD 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62885AE Supplier Thermo Scientific Chemicals Supplier No. J62885AE
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mL
Format Liquid
Product Type Cell Lysis Reagent
Concentration 5X
Content And Storage 2° to 8°C
What are the concentrations of the components in RIPA buffer with Triton X-100 (5X) (Cat. No. J62885.xx)?

RIPA buffer with Triton X-100 (5X) contains 250 mM Tris-HCl (pH 7.4), 750 mM NaCl, 5% Triton X-100, 2.5% sodium deoxycholate, and 0.5% SDS.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.