Learn More
Enzo Life Sciences RIPA cell lysis buffer 3, (100ml)
SDP
Catalog No. 502006819
Encompass_Preferred
Formulation: Liquid. 50mM TRIS-hydrogen chloride, pH 7.4, containing 150mM sodium chloride, 1mM EDTA, 1mM EGTA, 1.2% Triton X-100, 0.5% sodium deoxycholate, and 0.1% SDS. Long Term Storage: +4°C.
Catalog No. 50-200-6819 Supplier Enzo Life Sciences Supplier No. ADI801496
Formulation: Liquid. 50mM TRIS-hydrogen chloride, pH 7.4, containing 150mM sodium chloride, 1mM EDTA, 1mM EGTA, 1.2% Triton X-100, 0.5% sodium deoxycholate, and 0.1% SDS. Long Term Storage: +4°C.