Learn More
Boston Bioproducts Inc RIPA Buffer (2X) - 250ml
Catalog No. NC9879776
Encompass
The 2X RIPA Buffer is used for rapid, efficient cell lysis and solubilization of proteins from adherent and suspension cultured mammalian cells and tissue samples. With the combined action of its ionic detergents, the RIPA buffer is extremely useful for disruption of all proteins within cellular compartments, including nuclear, membrane, and mitochondrial. This buffer is compatible with many downstream applications, such as SDS-PAGE, immunoprecipitation, Western blot, and ELISA.
Catalog No. NC9879776 Supplier Boston Bioproducts Inc Supplier No. BP115X250ML
The 2X RIPA Buffer is used for rapid, efficient cell lysis and solubilization of proteins from adherent and suspension cultured mammalian cells and tissue samples. With the combined action of its ionic detergents, the RIPA buffer is extremely useful for disruption of all proteins within cellular compartments, including nuclear, membrane, and mitochondrial. This buffer is compatible with many downstream applications, such as SDS-PAGE, immunoprecipitation, Western blot, and ELISA.