Learn More
Santa Cruz Biotechnology RIPA Lysis Buffer System, 500 mL
Catalog No. NC0682466
Encompass
RIPA (Radioimmunoprecipitation) Lysis Buffer System is used to lyse cells and tissue, for radio immunoprecipitation assay (RIPA). RIPA lysis buffer has stronger denaturing capabilities than NP-40 or Triton X-100 and is particularly useful for disruption of nuclear membranes in the preparation of nuclear extracts. RIPA Lysis Buffer System includes protease inhibitors, making it ready for use in mammalian cell and tissue lysis.
Catalog No. NC0682466 Supplier Santa Cruz Biotechnology Supplier No. SC24948A
RIPA (Radioimmunoprecipitation) Lysis Buffer System is used to lyse cells and tissue, for radio immunoprecipitation assay (RIPA). RIPA lysis buffer has stronger denaturing capabilities than NP-40 or Triton X-100 and is particularly useful for disruption of nuclear membranes in the preparation of nuclear extracts. RIPA Lysis Buffer System includes protease inhibitors, making it ready for use in mammalian cell and tissue lysis.