Learn More
Biotang Inc RIPA WITH GLYCEROL(2X) 125 ML, BP-409, 100 mM Tris-HCl, 300 mM NaCl, 2% NP-40, 1% sodium deoxycholate, 0.2% SDS, and 10% Glycerol. pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1-micron filter.
Catalog No. 50155945 Shop All BIOTANG Inc Products
Encompass_Preferred
RIPA WITH GLYCEROL(2X) 125 ML, BP-409, 100 mM Tris-HCl, 300 mM NaCl, 2% NP-40, 1% sodium deoxycholate, 0.2% SDS, and 10% Glycerol. pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1-micron filter.
Catalog No. 50-155-945 Supplier Biotang Inc Supplier No. BTBB665
RIPA WITH GLYCEROL(2X) 125 ML, BP-409, 100 mM Tris-HCl, 300 mM NaCl, 2% NP-40, 1% sodium deoxycholate, 0.2% SDS, and 10% Glycerol. pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1-micron filter.