Learn More
Medchemexpress LLC HY-15685 5mg Medchemexpress, Ripasudil CAS:887375-67-9 Purity:>98%
SDP
Catalog No. 502029156
Encompass_Preferred
Medchemexpress, HY-15685 5mg Ripasudil CAS:887375-67-9 Ripasudil (K-115) is a specific inhibitor of ROCK, with IC50s of 19 and 51 nM for ROCK2 and ROCK1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9156 Supplier Medchemexpress LLC Supplier No. HY156855MG
Medchemexpress, HY-15685 5mg Ripasudil CAS:887375-67-9 Ripasudil (K-115) is a specific inhibitor of ROCK, with IC50s of 19 and 51 nM for ROCK2 and ROCK1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.