Learn More
Cayman Chemical Ripasudil (hydrochloride hydrate), Purity: ≥98%, CAS: 887375-67-9, 10 mg
SDP
Catalog No. 502984827
Encompass_Preferred
A ROCK1 and ROCK2 inhibitor (IC50s = 51 and 19 nM, respectively); selective for ROCK1 and ROCK2 over PKACα and PKC (IC50s = 2,100 and >10,000 nM, respectively); reduces intraocular pressure in cynomolgus monkey at 0.4%; increases the outflow facility of the aqueous humor in rabbit eyes at 0.4%; decreases intraocular pressure, retinal ganglion cell death, and thinning of inner retinal layers, as well as improves retinal function, in an EAAT1 knockout mouse model of normal tension glaucoma
Catalog No. 50-298-4827 Supplier Cayman Chemical Supplier No. 1992010 MG
A ROCK1 and ROCK2 inhibitor (IC50s = 51 and 19 nM, respectively); selective for ROCK1 and ROCK2 over PKACα and PKC (IC50s = 2,100 and >10,000 nM, respectively); reduces intraocular pressure in cynomolgus monkey at 0.4%; increases the outflow facility of the aqueous humor in rabbit eyes at 0.4%; decreases intraocular pressure, retinal ganglion cell death, and thinning of inner retinal layers, as well as improves retinal function, in an EAAT1 knockout mouse model of normal tension glaucoma