Learn More
Lascar Electronics Inc Pack of 10 Ripening Goods Temp & RH Logge
SDP
Supplier:  Lascar Electronics Inc ELCC2003 PK10
Encompass_Preferred
Available in packs of 10, the ultra low cost single-use EL-CC-2-003 PK10 Ripening Goods data logger measures and stores both temperature and humidity information. The unit comes pre-configured with High 14°C, Low 12°C Alarms - simply press the button to start logging. During a logging session, pressing the button will light up the status LEDs to give a quick indication of whether the temperature has exceeded an alarm condition at any point. Once your session is completed, tear open the IP67-rated plastic wallet to reveal the USB connector and plug it in to your PC. Use the free EasyLog CC software for Windows to download and graph the data.Please Note Whilst in this version of the logger temperature alarms are fixed, and humidity alarms are not set, Lascar can also provide custom configurations of this logger where both temperature and humidity alarms are set to your specifications. Please contact our Sales Team if this is of interest.
Catalog No. NC1856689