Learn More
Lascar Electronics Inc Pack of 10 Ripening Goods Data Logger
SDP
Catalog No. NC1856698
Encompass_Preferred
Sold in packs of 10 data loggers, the low cost EL-PDF-1-003 data logger comes with pre-configured alarms for use with Ripening Goods – simply press the Start button to begin logging. The LEDs give a quick status update & show whether the temperature has exceeded an alarm condition at any point. When a logging session is finished, simply tear open the waterproof wallet to reveal the USB connector & plug into your PC. The logger will then automatically generate a PDF report that you can save to your PC. The "Plug & Play" functionality & compact, lightweight design of this logger make it ideal for logistics use.This is designed to trigger if the temperature has been within a specified range for a specified amount of time through the duration of the whole log. There will be an LED indication when you are in the cumulative alarm region (both LEDs flash) but an alarm will not be triggered until the specified amount of time has been exceeded.
Catalog No. NC1856698 Supplier Lascar Electronics Inc Supplier No. ELPDF1003 PK10
Sold in packs of 10 data loggers, the low cost EL-PDF-1-003 data logger comes with pre-configured alarms for use with Ripening Goods – simply press the Start button to begin logging. The LEDs give a quick status update & show whether the temperature has exceeded an alarm condition at any point. When a logging session is finished, simply tear open the waterproof wallet to reveal the USB connector & plug into your PC. The logger will then automatically generate a PDF report that you can save to your PC. The "Plug & Play" functionality & compact, lightweight design of this logger make it ideal for logistics use.This is designed to trigger if the temperature has been within a specified range for a specified amount of time through the duration of the whole log. There will be an LED indication when you are in the cumulative alarm region (both LEDs flash) but an alarm will not be triggered until the specified amount of time has been exceeded.