Learn More
TARGETMOL CHEMICALS INC RIPISARTAN 5MG
SDP
Catalog No. 502580505
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Ripisartan (UP-269-6) is a potent and specific angiotensin II receptor antagonist that inhibits angiotensin II-mediated sympathetic tachycardia responses. purity: 98%
Catalog No. 50-258-0505 Supplier TARGETMOL CHEMICALS INC Supplier No. T285405MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Ripisartan (UP-269-6) is a potent and specific angiotensin II receptor antagonist that inhibits angiotensin II-mediated sympathetic tachycardia responses. purity: 98%