Learn More
Sino Biological Recombinant Human RET Protein (His Tag), Active
Catalog No. 502573447
Encompass_Preferred
Recombinant human RET (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-3447 Supplier Sino Biological Supplier No. R0211H50
Recombinant human RET (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.