Learn More
Sino Biological Recombinant Human RIOK2 Protein (Full Length, GST Tag)
Catalog No. 502573363
Encompass_Preferred
Recombinant full length human RIOK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3363 Supplier Sino Biological Supplier No. R0534G100
Recombinant full length human RIOK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.