Learn More
Aviva Systems Biology Corp RIPK1 Antibody - middle region (AVARP02038_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02038_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIPK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RIPK1. Alternate names: RIP, RIP1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-54084