Learn More
Medchemexpress LLC Ripk1-in-10 | 2682889-51-4 | 98.25% | C30H28F2N6O4 | 100 MG
SDP
Catalog No. 5000355513
Encompass_Preferred
RIPK1-IN-10 is a potent RIPK1 inhibitor, identified as example 37, extracted from patent WO2021160109. This product is intended for research use only and is not sold to patients. It is available as a solid, with a color ranging from white to light yellow.
  • Potent RIPK1 inhibitor
  • Intended for research use only
  • Available as a solid
  • Color ranges from white to light yellow
Catalog No. 50-003-55513 Supplier Medchemexpress LLC Supplier No. HY143728100MG
RIPK1-IN-10 is a potent RIPK1 inhibitor, identified as example 37, extracted from patent WO2021160109. This product is intended for research use only and is not sold to patients. It is available as a solid, with a color ranging from white to light yellow.
  • Potent RIPK1 inhibitor
  • Intended for research use only
  • Available as a solid
  • Color ranges from white to light yellow