Learn More
Medchemexpress LLC RIPK1-IN-4 | 1481641-08-0 | 98.89% | 401.46 | 1 ML
SDP
Catalog No. 5000385270
Encompass_Preferred
RIPK1-IN-4 (compound 8) is identified as a potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase. It binds to a DLG-out inactive form of RIP1 with IC50s of 16 nM and 10 nM for RIP1 and ADP-Glo kinase, respectively. It is intended for research use only.
  • Potent and selective type II kinase inhibitor of RIP1 kinase.
  • Binds to a DLG-out inactive form of RIP1.
  • High purity: 98.89%.
  • Available in solid and solution forms for various research needs.
  • Detailed solubility information and protocols for in vitro and in vivo applications are provided.
Catalog No. 50-003-85270 Supplier Medchemexpress LLC Supplier No. HY1890110MM/1ML
Add to Cart
Add to Cart
RIPK1-IN-4 (compound 8) is identified as a potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase. It binds to a DLG-out inactive form of RIP1 with IC50s of 16 nM and 10 nM for RIP1 and ADP-Glo kinase, respectively. It is intended for research use only.
  • Potent and selective type II kinase inhibitor of RIP1 kinase.
  • Binds to a DLG-out inactive form of RIP1.
  • High purity: 98.89%.
  • Available in solid and solution forms for various research needs.
  • Detailed solubility information and protocols for in vitro and in vivo applications are provided.