Learn More
Medchemexpress LLC RIPK1-IN-4 | 1481641-08-0 | 98.9% | 401.46 | C23H23N5O2 | 10 MG
SDP
Catalog No. 5000298311
Encompass_Preferred
A potent, selective type II kinase inhibitor of receptor-interacting protein 1 (RIP1) that binds the DLG-out inactive conformation. It exhibits low-nanomolar biochemical potency (RIP1 IC50 16 nM; ADP-Glo IC50 10 nM) and shows cellular activity against RIP1-mediated necroptosis, making it suitable as a research tool for studies of RIP1 signaling and necroptosis.
  • Potent biochemical inhibition of RIP1 (IC50 16 nM).
  • Low-nanomolar ADP-Glo kinase activity (IC50 10 nM).
  • Cellular efficacy in necroptosis models (L929, U-937).
  • High purity and white to off-white solid appearance.
  • High DMSO solubility (250 mg/mL) with ultrasonic assistance.
  • Stable when stored per recommended conditions (powder and solvent).
Catalog No. 50-002-98311 Supplier Medchemexpress LLC Supplier No. HY1890110MG
A potent, selective type II kinase inhibitor of receptor-interacting protein 1 (RIP1) that binds the DLG-out inactive conformation. It exhibits low-nanomolar biochemical potency (RIP1 IC50 16 nM; ADP-Glo IC50 10 nM) and shows cellular activity against RIP1-mediated necroptosis, making it suitable as a research tool for studies of RIP1 signaling and necroptosis.
  • Potent biochemical inhibition of RIP1 (IC50 16 nM).
  • Low-nanomolar ADP-Glo kinase activity (IC50 10 nM).
  • Cellular efficacy in necroptosis models (L929, U-937).
  • High purity and white to off-white solid appearance.
  • High DMSO solubility (250 mg/mL) with ultrasonic assistance.
  • Stable when stored per recommended conditions (powder and solvent).