Learn More
Medchemexpress LLC HY-18901 5mg Medchemexpress, RIPK1-IN-4 CAS:1481641-08-0 Purity:>98%
SDP
Catalog No. 501970016
Encompass_Preferred
Medchemexpress, HY-18901 5mg RIPK1-IN-4 CAS:1481641-08-0 RIPK1-IN-4 (compound 8) is a potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase and binds to a DLG-out inactive form of RIP1 with an IC50s of 16 nM and 10 nM for RIP1 and ADP-Glo kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0016 Supplier Medchemexpress LLC Supplier No. HY189015MG
Medchemexpress, HY-18901 5mg RIPK1-IN-4 CAS:1481641-08-0 RIPK1-IN-4 (compound 8) is a potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase and binds to a DLG-out inactive form of RIP1 with an IC50s of 16 nM and 10 nM for RIP1 and ADP-Glo kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.