Learn More
Medchemexpress LLC HY-119933 10mg Medchemexpress, RIPK1-IN-7 CAS:2300982-44-7 Purity:>98%
SDP
Catalog No. 501969554
Encompass_Preferred
Medchemexpress, HY-119933 10mg RIPK1-IN-7 CAS:2300982-44-7 RIPK1-IN-7 is a potent and selective RIPK1 inhibitor with a Kd of 4 nM and an enzymatic IC50 of 11 nM. RIPK1-IN-7 exhibits excellent antimetastasis activity in the experimental B16 melanoma lung metastasis model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9554 Supplier Medchemexpress LLC Supplier No. HY11993310MG
Medchemexpress, HY-119933 10mg RIPK1-IN-7 CAS:2300982-44-7 RIPK1-IN-7 is a potent and selective RIPK1 inhibitor with a Kd of 4 nM and an enzymatic IC50 of 11 nM. RIPK1-IN-7 exhibits excellent antimetastasis activity in the experimental B16 melanoma lung metastasis model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.