Learn More
Aviva Systems Biology Corp RIPK2 Antibody - middle region (AVARP02047_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02047_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIPK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RIPK2. Alternate names: CARD3, CARDIAK, CCK, GIG30, RICK, RIP2ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62490